Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KGF Goat anti-Human, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA124683

Catalog No. PIPA124683


Add to cart

Description

Description

PA1-24683 detects KGF from human samples. PA1-24683 has been successfully used in neutralization, inhibition assays, and Western blot procedures. The Neutralization Dose 50 (ND50) for anti-human FGF7 (KGF) is approximately 6 to 12 µg/mL in the presence of 125 ng/mL of recombit human FGF7 using the human 4MBr-5 cell line. The PA1-24683 immunogen is recombit, full length human protein. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

KGF
Polyclonal
Unconjugated
FGF7
Liquid
Goat
IgG
50 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
2252
ELISA, Inhibition Assays, Neutralization, Western Blot
0.25 mg/mL
PBS with no preservative
P21781
FGF7
Recombit, full length human protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit