Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Ki-67/MKI67, FITC, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NB11090592F

Catalog No. NB11090592F

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Ki-67/MKI67 Polyclonal antibody specifically detects Ki-67/MKI67 in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

Ki-67/MKI67
1mg/mL
Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
P46013
antigen identified by monoclonal Ki-67, antigen KI-67, Ki67, Ki-67, KIA, Marker Of Proliferation Ki-67, MIB-1, PPP1R105, proliferation-related Ki-67 antigen, Protein Phosphatase 1, Regulatory Subunit 105
Rabbit
IgG
0.05 ml
Cell Biology, Cell Cycle and Replication, Cellular Markers, Growth and Development, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Stem Cell Markers
Primary
4288
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
FITC
PBS with 0.05% Sodium Azide
Affinity Purified
MKI67
Synthetic peptide made to an internal portion of the human Ki67 protein (within residues 1200-1300). [Swiss-Prot: P46013]
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
Human, Mouse
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only