Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Ki-67/MKI67, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Specifications

Host Species Rabbit
Isotype IgG
Immunogen Synthetic peptide made to an internal portion of the human Ki67 protein (within residues 2100-2200). [Swiss-Prot# P46013]
Antigen Ki-67/MKI67
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB11090593
View Documents
Novus Biologicals
NB11090593
0.05 ml This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Ki-67/MKI67 Polyclonal antibody specifically detects Ki-67/MKI67 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Rabbit
Synthetic peptide made to an internal portion of the human Ki67 protein (within residues 2100-2200). [Swiss-Prot# P46013]
Polyclonal
Primary
Immunohistochemistry, Immunohistochemistry (Paraffin)
4288
1 mg/mL
IgG
Ki-67/MKI67
Affinity Purified
RUO
P46013
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.1% Sodium Azide
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit