Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-KIAA0494, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP186098
Description
EFCAB14 Polyclonal antibody specifically detects EFCAB14 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
KIAA0494 | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
hypothetical protein LOC9813, KIAA0494, RP11-8J9.3 | |
Rabbit | |
IgG | |
0.1 ml | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
Polyclonal | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200-1:500 | |
Affinity Purified | |
EFCAB14 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:NKVWINITEMNKQISLLTSAVNHLKANVKSAADLISLPTTVEGLQKSVASIGNTLNSVHLAVEALQKTVDEHKKTMELLQSDMNQHFLKETPGSNQIIPSPSATSELDNKTHSENLKQDILYLHNSLEEVNSALV | |
Immunogen affinity purified | |
RUO | |
Primary | |
9813 | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only