Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIAA1310 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524238

Catalog No. PIPA524238

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

KIAA1310
Polyclonal
PBS with 0.09% sodium azide
Q9P2N6
KIAA1310, NSL3, Rcd1, serum inhibited-related protein, testis development protein PRTD, uncharacterized protein KIAA1310
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55683
Immunohistochemistry, Western Blot
Unconjugated
KANSL3
Liquid
KANSL3
KLH conjugated synthetic peptide between 398-427 amino acids from the central region of human KIAA1310
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit