Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIAA1688 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525859

Catalog No. PIPA525859

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

KIAA1688
Polyclonal
PBS with 0.09% sodium azide
Q9C0H5, P59281
CrGAP, Vilse, RhoGAP93B homolog, crossGAP homolog, rho GTPase-activating protein 39
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
223666, 80728
Western Blot
Unconjugated
ARHGAP39
Liquid
ARHGAP39
KLH conjugated synthetic peptide between 852-880 amino acids from the C-terminal region of human KIAA1688
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit