Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIF14 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA527859
Description
Recommended positive controls: Jurkat, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
KIF14 Polyclonal antibody specifically detects KIF14 in Human samples. It is validated for Western Blot.Specifications
KIF14 | |
Polyclonal | |
Unconjugated | |
KIF14 | |
D1Ertd367e; E130203M01; KIAA0042; KIF14; kinesin family member 14; kinesin-like protein KIF14; MKS12; N-3 kinesin; truncated kinesin family member 14 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9928 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q15058 | |
KIF14 | |
Recombinant fragment corresponding to a region within amino acids 35 and 323 of Human KIF14 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction