Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIF3C Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA514373

Catalog No. PIPA514373

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

KIF3C is a microtubule-based anterograde translocator for membranous organelles.
Specifications

Specifications

KIF3C
Polyclonal
PBS with 0.09% sodium azide
O14782
Kinesin-like protein KIF3C ; KIF3C
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3797
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
KIF3C
Liquid
KIF3C
KLH conjugated synthetic peptide between 713-741 amino acids from the C-terminal region of human KIF3C
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit