Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIF4A Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA528142

Catalog No. PIPA528142


Add to cart

Description

Description

PA5-28142 targets KIF4A in WB applications and shows reactivity with Human samples. The PA5-28142 immunogen is synthetic peptide corresponding to a region within amino acids 1169 and 1232 of Human KIF4A. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Kinesins, such as KIF4A, are microtubule-based motor proteins that generate directional movement along microtubules. They are involved in many crucial cellular processes, including cell division (Zhu and Jiang, 2005 [PubMed 15625105]).
Specifications

Specifications

KIF4A
Polyclonal
Unconjugated
KIF4A
Liquid
KIF4A
Synthetic peptide corresponding to a region within amino acids 1169 and 1232 of Human KIF4A
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 10% glycerol and 0.01% thimerosal; pH 7
O95239
KIF4, KIF4G1, chromokinesin-A, chromosome-associated kinesin KIF4A
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
24137
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit