Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIF7 Mouse anti-Human, Mouse, Rat, Clone: 3F8, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA518325

Catalog No. PIMA518325


Add to cart

Description

Description

Do not aliquot.

This gene encodes a cilia-associated protein belonging to the kinesin family. This protein plays a role in the sonic hedgehog (SHH) signaling pathway through the regulation of GLI transcription factors. It functions as a negative regulator of the SHH pathway by preventing inappropriate activation of GLI2 in the absence of ligand, and as a positive regulator by preventing the processing of GLI3 into its repressor form. Mutations in this gene have been associated with various ciliopathies. [provided by RefSeq, Oct 2011].
Specifications

Specifications

KIF7
Monoclonal
1 mg/mL
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Q2M1P5, B7ZNG0
KIF7
A synthetic peptide derived from human Kif 7
Affinity chromatography
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Immunohistochemistry (Paraffin)
3F8
Unconjugated
KIF7
Liquid
Mouse
IgG1
100 μg
-20°C
Primary
16576, 293047, 374654
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit