Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIF9 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA525846
Description
Specifications
KIF9 | |
Polyclonal | |
KIF9 | |
kinesin protein 9, kinesin-like protein KIF9 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16578, 64147 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Q9HAQ2, Q9WV04 | |
KIF9 | |
KLH conjugated synthetic peptide between 581-609 amino acids from the C-terminal region of human KIF9 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction