Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Kilon/NEGR1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP132529

Catalog No. NBP132529


Add to cart

Description

Description

Kilon/NEGR1 Polyclonal antibody specifically detects Kilon/NEGR1 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Kilon/NEGR1
0.3mg/mL
Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Affinity Purified
NEGR1
Recombinant protein encompassing a sequence within the center region of human NEGR1. The exact sequence is proprietary.
39 kDa
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in Mouse. Not yet tested in any other species.
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
a kindred of IgLON, IgLON family member 4, IGLON4DMML2433, KILONMGC46680, neuronal growth regulator 1, neurotractin, Ntra
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
257194
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only