Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-kinectin, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232262

Catalog No. NBP232262


Add to cart

Description

Description

kinectin Polyclonal antibody specifically detects kinectin in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

kinectin
1.0 mg/mL
Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg lysate
NP_001072989.1
kinectin, kinectin 1 (kinesin receptor), MGC133337
Rabbit
IgG
100 ul
Store at 4C. Do not freeze.
Polyclonal
Human
Western Blot, Immunoprecipitation
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
KTN1
The immunogen this antibody was made to, maps to a region between residue 1307 to 1357 of human Kinectin using the numbering given in entry NP_001072989.1 (GeneID 3895).
Immunogen affinity purified
RUO
Primary
3895
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only