Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIF14, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA126592

Catalog No. PIPA126592

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

PA1-26592 detects KIF14 from human samples.PA1-26592 has been successfully used in Western blot procedures.The PA1-26592 immunogen is a synthetic peptide corresponding to a portion of human Kinesin Family Member 14 (KIF 14) encoded within exon 16.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Kinesin 14
Polyclonal
Unconjugated
Q15058
Rabbit
Antigen affinity chromatography
RUO
9928
Store at 4°C, do not freeze
Purified
Immunoprecipitation, Western Blot
1mg/mL
KIF14
KIF14
Synthetic peptide corresponding to a portion of human Kinesin Family Member 14 (KIF 14) encoded within exon 16.
50 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit