Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIFC2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA1641
Description
PA1-641 detects kinesin C2 (KIFC2) from mouse tissues. PA1-641 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ∽100 kDa protein representing KIFC2 from mouse brain homogenate. PA1-641 immunizing peptide corresponds to amino acid residues 780-792 from mouse KIFC2. PA1-641 immunizing peptide (Cat. No. PEP-041) is available for use in neutralization and control experiments.
Kinesins are a superfamily of microtubule-associated motor proteins involved in a variety of cellular processes including membranous organelle transport and cell division. Kinesin has been found in a variety of organisms and cell types and is subject to spatial and temporal regulation. These proteins have a modular structure which includes a conserved motor domain of approximately 350 amino acids, which is responsible for microtubule binding and ATP hydrolysis. In addition to the motor domain, subfamily members share common domain organization, exhibit sequence similarity, motility properties, and cellular functions outside of the motor domain. An expansive phylogenetic tree of kinesins and kinesin-related proteins (KRPs) has been assembled based on this information. Kinesin C2 (KIFC2) is a C-type kinesin based on the position of the motor domain towards the carboxyl-terminus of the protein and is similar to members of the karyogamy protein 3 (KAR3) family involved in cell division. KIFC2 protein expression has been demonstrated in axons and dendrites of the central and peripheral nervous system. Unlike other C-type motor proteins, KIFC2 appears to play a role in retrograde axonal transport.Specifications
KIFC2 | |
Polyclonal | |
Unconjugated | |
O08672 | |
KIFC2 | |
Synthetic Peptide: C S(780) G L T L E P P G D P P P(792) | |
200 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Western Blot | |
1 mg/mL | |
Kifc2 | |
KIFC2, Kinesin family member C2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16581 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction