Learn More
anti-KIR2DL1/CD158a, Fluorescein, Clone: 143211, R&D Systems™
Manufacturer:  R&D Systems FAB1844F025
 View more versions of this product
Catalog No. FAB1844F025
Add to cart
Description
Description
Applications:
Flow Cytometry
Specifications
Specifications
KIR2DL1/KIR2DS5
143211
Flow Cytometry 10 uL/10^6 cells
P43626
CD158a, CD158g, NKAT9
Mouse
IgG1
25 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects human KIR2DL1/KIR2DS5 on transfected cells. It does not stain cells transfected with KIR2DL2, 2DL3, 2DL4, 2DL5, 2DS1, 2DS2, 2DS4, 3DL1, 3DL2, or 3DS1.
Flow Cytometry
Fluorescein
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
KIR2DL1
BaF3 mouse pro-B cell line transfected with human KIR2DL1/KIR2DS5 Accession # P43626
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3802
Human