Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIR2DL2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525666

Catalog No. PIPA525666


Add to cart

Description

Description

Killer cell immunoglobulin-like receptors (KIRs) are transmembrane glycoproteins expressed by natural killer cells and subsets of T cells. The KIR genes are polymorphic and highly homologous and they are found in a cluster on chromosome 19q13.4 within the 1 Mb leukocyte receptor complex (LRC). The gene content of the KIR gene cluster varies among haplotypes, although several  ft.framework ft. genes are found in all haplotypes (KIR3DL3, KIR3DP1, KIR3DL4, KIR3DL2). The KIR proteins are classified by the number of extracellular immunoglobulin domains (2D or 3D) and by whether they have a long (L) or short (S) cytoplasmic domain. KIR proteins with the long cytoplasmic domain transduce inhibitory signals upon ligand binding via an immune tyrosine-based inhibitory motif (ITIM), while KIR proteins with the short cytoplasmic domain lack the ITIM motif and instead associate with the TYRO protein tyrosine kinase binding protein to transduce activating signals. The ligands for several KIR proteins are subsets of HLA class I molecules; thus, KIR proteins are thought to play an important role in regulation of the immune response.
Specifications

Specifications

KIR2DL2
Polyclonal
PBS with 0.09% sodium azide
P43627
CD158B1, CD158b, NKAT6, p58.2, CD158 antigen-like family member B1, MHC class I NK cell receptor, NKAT-6, killer cell immunoglobulin-like receptor 2DL2, natural killer-associated transcript 6, p58 NK receptor CL-43, p58 natural killer cell receptor clone CL-43
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3803
Flow Cytometry, Western Blot
Unconjugated
KIR2DL2
Liquid
KIR2DL2
KLH conjugated synthetic peptide between 263-291 amino acids from the C-terminal region of human KIR2DL2
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit