Learn More
anti-KIR2DL3/CD158b2, Fluorescein, Clone: 180701, R&D Systems™
Manufacturer:  R&D Systems FAB2014F025
 View more versions of this product
Catalog No. FAB2014F025
Add to cart
Description
Description
Applications:
Flow Cytometry
Specifications
Specifications
KIR2DL3/CD158b2
180701
Flow Cytometry 10 uL/10^6 cells
P43628
CD158 antigen-like family member B2, CD158b2, CD158b2 antigen, CD158B2CD158b, cl-6, killer cell immunoglobulin-like receptor 2DL3, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 3, Killer inhibitory receptor cl 2-3, KIR-023GB, KIRCL23, KIR-K7b, KIR-K7c, MGC129943, MHC class I NK cell receptor, natural killer associated transcript 2, natural killer cell inhibitory receptor KIR2DL3, Natural killer-associated transcript 2, NKAT, nkat2, NKAT-2, nkat2a, NKAT2B, NKAT2GL183, NK-receptor, p58, p58 natural killer cell receptor clone CL-6, p58 NK receptor CL-6, p58.2 MHC class-I specific NK receptor, p58.2 MHC class-I-specific NK receptor
Mouse
IgG2a
25 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects human KIR2DL3/CD158b2. Stains BaF3 cells transfected with human KIR2DL3 in flow cytometry. Does not stain BaF3 cells transfected with KIR2DL1, 2DL2, 2DL4, 2DL5, 2DS1, 2DS2, 2DS4, 3DL1, 3DL2, or 3DS1.
Flow Cytometry
Fluorescein
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
KIR2DL3
BaF3 mouse pro-B cell line transfected with human KIR2DL3/CD158b2 Accession # P43628
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3804
Human