Learn More
anti-KIR2DL4/CD158d, PE, Clone: 181703, R&D Systems™
Manufacturer:  R&D Systems FAB2238P025
 View more versions of this product
Catalog No. FAB2238P025
Add to cart
Description
Description
Applications:
Flow Cytometry
Specifications
Specifications
KIR2DL4/CD158d
181703
Flow Cytometry 10 uL/10^6 cells
Q99706
103AS, 15.212, CD158 antigen-like family member D, CD158d, CD158d antigen, CD158DKIR103ASKiller cell inhibitory receptor 103AS, G9P, killer cell immunoglobulin-like receptor 2DL4, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 4, killer Ig receptor, KIR103, KIR-103AS, MHC class I NK cell receptor KIR103AS, natural killer cell inhibitory receptor, NK cell receptor
Mouse
IgG2a
25 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Stains human KIR2DL4 transfected BaF/3 cells. It does not recognize transfectants that express KIR2DL1, 2DL2, 2DL3, 2DL5, 2DS1, 2DS2, 2DS4, 3DL1, 3DL2, or 3DS1.
Flow Cytometry
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
KIR2DL4
BaF3 mouse pro-B cell line transfected with human KIR2DL4, Accession # Q99706
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3805
Human