Learn More
anti-KIR3DL2/CD158k, Alexa Fluor 700, Clone: 539304, R&D Systems™
Manufacturer:  R&D Systems FAB2878N100
 View more versions of this product
Catalog No. FAB2878N100
Add to cart
Description
Description
Applications:
Flow Cytometry
Specifications
Specifications
KIR3DL2/CD158k
539304
Flow Cytometry 5 uL/10^6 cells
P43630
killer cell immunoglobulin-like receptor 3DL2, CD158K, CL-5, killer cell immunoglobulin-like receptor, three domains, long cytoplasmic tail, 2, NKAT4, NKAT-4, NKAT4A, NKAT4B, p140
Mouse
IgG1
100 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects human KIR3DL2/CD158k in direct ELISAs.
Flow Cytometry
Alexa Fluor 700
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
KIR3DL2
BaF3 mouse pro-B cell line transfected with human KIR3DL2/CD158k Accession # P43630
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3812
Human