Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIR3DL3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526178

Catalog No. PIPA526178


Add to cart

Description

Description

KIR3DL3 is transmembrane glycoproteins expressed by natural killer cells and subsets of T cells. This protein are polymorphic and highly homologous and they are found in a cluster on chromosome 19q13.4 within the 1 Mb leukocyte receptor complex (LRC).
Specifications

Specifications

KIR3DL3
Polyclonal
PBS with 0.09% sodium azide
Q8N743
CD158Z, KIR3DL7, KIR44, KIRC1, CD158 antigen-like family member Z, killer cell Ig-like receptor KIR3DL7, killer cell immunoglobulin-like receptor 3DL3, killer cell inhibitory receptor 1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
115653
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
KIR3DL3
Liquid
KIR3DL3
KLH conjugated synthetic peptide between 132-158 amino acids from the central region of human KIR3DL3
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit