Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLHDC2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524748

Catalog No. PIPA524748


Add to cart

Description

Description

KLDC2 represses CREB3-mediated transcription by interfering with CREB3-DNA binding.
Specifications

Specifications

KLHDC2
Polyclonal
Unconjugated
KLHDC2
Liquid
KLHDC2
KLH conjugated synthetic peptide between 293-320 amino acids from the C-terminal region of human KLDC2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q9Y2U9, Q4G5Y1
HCLP-1, LCP, hepatocellular carcinoma-associated antigen 33, host cell factor homolog LCP, host cell factor-like protein 1, kelch domain-containing protein 2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
23588, 69554
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit