Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Klebsiella, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA128993

Catalog No. PIPA128993

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Klebsiella are non-motile, rod-shaped, aerobic bacteria that possess a prominent polysaccharide capsule. Some of these bacteria produce an extracellular toxic complex that has been shown to be lethal for and produce extensive lung pathology in mice. It is composed of 63% capsular polysaccharide, 30% lipopolysaccharide, and 7% protein; when introduced to experimental animals in sublethal doses, the animals built up immunization due to antibody production. Klebsiella pneumoniae is one of the most common Gram-negative bacteria seen by physicians worldwide. Pneumonias that are caused by Klebsiella pneumoniae are difficult to control; mortality rates have even been reported as up to 50% after antibiotic treatment. Nitrogen metabolism in Klebsiella aerogenes has also been studied throroughly.

PA1-28993 detects Klebsiella from bacteria samples.PA1-28993 has been successfully used in immunofluorescence applications.The PA1-28993 immunogen is: Klebsiella pneumoniae, ATCC #43816.Store at 4ºC short term. For extended storage, aliquot and store at -20ºC or below. Avoid freeze/thaw cycles.
Specifications

Specifications

Klebsiella
Polyclonal
Unconjugated
Purified
Klebsiella pneumoniae, ATCC #43816.
Protein A
RUO
Antibody
Polyclonal
Immunocytochemistry, Immunofluorescence
4mg/mL
PBS with 0.1% sodium azide; pH 7.2
Rabbit
IgG
100μL
Store at -20°C
Primary
Bacteria
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit