Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLF10 Mouse anti-Human, Clone: 3E11, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA520125

Catalog No. PIMA520125


Add to cart

Description

Description

Peptide Sequence: LSDTAKPHIA APFKEEEKSP VSAPKLPKAQ ATSVIRHTAD AQLCNHQTCP MKAASILNYQ NNSFRRRTHL NVEAARKNIP CAAVSPNRSK CERNTVADVD

KLF10 is a nuclear protein belonging to the Sp1 C2H2-type zinc-finger protein family with three C2H2-type zinc fingers. KLF10 is a transcriptional repressor involved in the regulation of cell growth and inhibits cell growth. It binds to the consensus sequence 5 ft.-GGTGTG-3 ft. and is induced by TGF-beta thus playing a role in linking TGF-beta-mediated signaling cascades to the regulation of pancreatic epithelial cell growth. KLF10 is involved in inducing and promoting apoptosis through the mitochondrial apoptotic pathway and may inhibit Smad7 expression in bronchial epithelial cells. Ubiquitous expression of KLF10 is seen in most of the tissues. Reports suggest that it may mediate up-regulation of TGFBI in VHL-deficient tumors and other cancers. Over expression of KLF10 may be associated with renal clear cell carcinoma and other tumors. KLF11 is a zinc finger transcription factor that binds to SP1-like sequences in epsilon- and gamma-globin gene promoters. This binding inhibits cell growth and causes apoptosis. Defects in this gene are a cause of maturity-onset diabetes of the young type 7 (MODY7). Three transcript variants encoding two different isoforms have been found for this gene.
Specifications

Specifications

KLF10
Monoclonal
1 mg/mL
PBS with no preservative; pH 7.4
Q13118
EGRA, TIEG, TIEG1, EGR-alpha; Krueppel-like factor 10; TGFB inducible early growth response; TGFB-inducible early growth response protein 1; TIEG-1; transforming growth factor-beta-inducible early growth response protein 1
Mouse
IgG2a, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
7071
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
3E11
Unconjugated
KLF10
Liquid
KLF10
KLF10 (AAH11538.1, 111 a.a. ∽ 210 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Protein A
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit