Learn More
KLF11 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA528031
Catalog No. PIPA528031
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
PA5-28032 targets RAB3B in WB applications and shows reactivity with Human samples. The PA5-28032 immunogen is recombit fragment corresponding to a region within amino acids 1 and 219 of Human RAB3B.
The protein encoded by this gene is a zinc finger transcription factor that binds to SP1-like sequences in epsilon- and gamma-globin gene promoters. This binding inhibits cell growth and causes apoptosis. Defects in this gene are a cause of maturity-onset diabetes of the young type 7 (MODY7). Three transcript variants encoding two different isoforms have been found for this gene.
Specifications
Specifications
KLF11
Polyclonal
Unconjugated
KLF11
Liquid
KLF11
Recombit fragment corresponding to a region within amino acids 149 and 408 of Human KLF11
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
O14901
FKLF, FKLF1, MODY7, TIEG2, Tieg3, Krueppel-like factor 11, TGFB-inducible early growth response protein 2, TIEG-2, transforming growth factor-beta-inducible early growth response protein 2
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
8462