Learn More
anti-KLF15, Polyclonal, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP130923
Catalog No. NBP130923
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
KLF15
1.0mg/mL
Western Blot 1:500-1:3000
Affinity Purified
KLF15
Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human KLF15. The exact sequence is proprietary.
44 kDa
100 ul
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.
Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
DKFZp779M1320, kidney-enriched Kruppel-like factor, KKLFKidney-enriched krueppel-like factor, Krueppel-like factor 15, Kruppel-like factor 15
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
28999
Human