Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLF17 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525421

Catalog No. PIPA525421

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

KLF17 binds G/C-rich sites via its zinc fingers and activates transcription from CACCC-box elements. It may be a germ cell-specific transcription factor that plays important roles in spermatid differentiation and oocyte development.
Specifications

Specifications

KLF17
Polyclonal
PBS with 0.09% sodium azide
Q5JT82
ZNF393, Zfp393, Krueppel-like factor 17, novel zinc-finger protein, zinc finger protein 393
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
128209
Flow Cytometry, Western Blot
Unconjugated
KLF17
Liquid
KLF17
KLH conjugated synthetic peptide between 243-271 amino acids from the C-terminal region of human KLF17
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit