Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLF6 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA513536

Catalog No. PIPA513536

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

COPEB is a member of the Kruppel-like family of transcription factors. The zinc finger protein is a transcriptional activator, and functions as a tumor suppressor.
Specifications

Specifications

KLF6
Polyclonal
PBS with 0.09% sodium azide
Q99612
Krueppel-like factor 6; Core promoter element-binding protein; B-cell-derived protein 1; Proto-oncogene BCD1; Transcription factor Zf9; GC-rich sites-binding factor GBF; ST12; CPBP; COPEB; BCD1; KLF6
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1316
Flow Cytometry, Western Blot
Unconjugated
KLF6
Liquid
KLF6
KLH conjugated synthetic peptide between 56-82 amino acids from the N-terminal region of human KLF6
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit