Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-KLH, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP130443

Catalog No. NBP130443


Add to cart

Description

Description

KLH Polyclonal antibody specifically detects KLH in Non-species specific samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

KLH
1.0 mg/mL
Western Blot 1:10000-1:100000, ELISA 1:10000-1:100000
Affinity Purified
Rabbit
IgG
1 ml
Store at 4C. Do not freeze.
Polyclonal
ELISA, Western Blot
Unconjugated
Phosphate Buffered Saline (PBS) containing 0.2%BSA with 0.09% Sodium Azide
keyhole limpet hemocyanin (KLH)
KLH was used as the immunogen.
Immunogen affinity purified
RUO
Primary
Non-species specific
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only