Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLHDC1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524341

Catalog No. PIPA524341


Add to cart

Description

Description

KLHDC1 contains 6 Kelch repeats and is widely expressed, with high levels in skeletal muscle, pancreas and liver. KLHDC1 and KLHDC2 have differential localization and activity in cultured mammalian cells. The exact function of KLHDC1 remains unknown.
Specifications

Specifications

KLHDC1
Polyclonal
PBS with 0.09% sodium azide
Q8N7A1, Q80YG3
MST025, kelch domain-containing protein 1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
122773, 271005
Immunohistochemistry, Western Blot
Unconjugated
KLHDC1
Liquid
KLHDC1
KLH conjugated synthetic peptide between 69-97 amino acids from the N-terminal region of human KLHDC1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit