Learn More
KLHL6 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA525693
Catalog No. PIPA525693
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Involved in B-lymphocyte antigen receptor signaling and germinal center formation.
Specifications
Specifications
KLHL6
Polyclonal
Unconjugated
KLHL6
Liquid
KLHL6
KLH conjugated synthetic peptide between 543-571 amino acids from the C-terminal region of human KLHL6
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Flow Cytometry, Immunohistochemistry, Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q8WZ60, Q6V595
kelch-like protein 6, kelch-like protein KLHL6
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
239743, 89857