Learn More
kallikrein-related peptidase 14, Mouse, Polyclonal Antibody, Abnova
Mouse polyclonal antibody raised against a full-length human KLK14 protein.
Manufacturer: Abnova Corporation H00043847B01
Description
Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in a cluster on chromosome 19. An additional transcript variant has been described but its full length nature has not been determined. [provided by RefSeq
Sequence: MFLLLTALQVLAIAMTQSQEDENKIIGGHTCTRSSQPWQAALLAGPRRRFLCGGALLSGQWVITAAHCGRPILQVALGKHNLRRWEATQQVLRVVRQVTHPNYNSRTHDNDLMLLQLQQPARIGRAVRPIEVTQACASPGTSCRVSGWGTISSPIARYPASLQCVNINISPDEVCQKAYPRTITPGMVCAGVPQGGKDSCQGDSGGPLVCRGQLQGLVSWGMERCALPGYPGVYTNLCKYRSWIEETMRDKSpecifications
kallikrein-related peptidase 14 | |
Unconjugated | |
No additive | |
ENST00000156499 | |
KLK-L6 | |
Mouse | |
50 uL | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Primary | |
43847 |
Western Blot | |
Mouse polyclonal antibody raised against a full-length human KLK14 protein. | |
KLK14 | |
Serum | |
KLK14 | |
KLK14 (ENSP00000156499, 1 a.a. ~ 251 a.a) full-length human protein. | |
RUO | |
Y | |
Polyclonal | |
Human |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.