Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLRA2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA520687
Description
A suggested positive control is mouse spleen tissue lysate. PA5-20687 can be used with blocking peptide PEP-0803.
KLRA2 Polyclonal antibody specifically detects KLRA2 in Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Specifications
KLRA2 | |
Polyclonal | |
Unconjugated | |
Klra2 | |
killer cell lectin-like receptor 2; killer cell lectin-like receptor subfamily A member 30; killer cell lectin-like receptor, subfamily A, member 2; Klra2; Klra30; Ly49 inhibitory receptor 6; Ly49b; ly-49b; Ly49i6; lymphocyte antigen 49b; membrane protein Ly49b; NK cell inhibitory receptor Klra2; T-cell surface glycoprotein Ly49b; T-cell surface glycoprotein Ly-49B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16633, 494193 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q60660 | |
Klra2 | |
A 14 amino acid peptide near the carboxy terminus of mouse KLRA2. | |
100 μg | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction