Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-KLRC1 Matched Antibody Pair, Abnova™

Manufacturer:  Abnova Corporation H00003821AP51

Catalog No. 89-023-492


Add to cart

Includes: Capture antibody: mouse monoclonal anti-KLRC1 (100µg); Detection antibody: rabbit purified polyclonal anti-KLRC1 (50µg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols

Description

Description

This antibody pair set comes with matched antibody pair to detect and quantify protein level of human KLRC1

Host Species: Mouse, Rabbit
Species Reactivity: Hu

ELISA Pair (Transfected Lysate)

Specifications

Specifications

KLRC1
Unconjugated
KLRC1
CD159A|MGC13374|MGC59791|NKG2|NKG2A
Murine, Rabbit
RUO
Primary
3821
Human
ELISA
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human KLRC1.
Purified
KLRC1
1 Set
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Monoclonal/Polyclonal
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-KLRC1 (100 ug) 2. Detection antibody: rabbit purified polyclonal anti-KLRC1 (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit