Learn More
KLRG1 Hamster anti-Mouse, Brilliant Violet 421, Clone: 2F1, BD
Hamster Monoclonal Antibody
Manufacturer:  BD Biosciences 562897
Catalog No. BDB562897
Add to cart
Description
Description
The 2F1 monoclonal antibody specifically binds to KLRG1 (Killer cell Lectin-like Receptor mmunoreceptor Tyrosine-based Inhibitory Motif);its ligand has not beenidentified KLRG1 is expressed mainly as a homodimeric molecule consisting of two N-glycosylated subunits of approximately 30-38 kDa. The level of KLRG1 expression is reduced in MHC class I-deficient mice, although direct binding of KLRG1 to MHC class I antigens could not be detected. Cross-linking of KLRG1 by 2F1 mAb reduces TCR-mediated Ca++mobilization and cytotoxic responses (but not IFN-γ production) by CD8+ T cellsand inhibits IFN-γ and TNF-αproduction and redirected lysis by NK cells.
Specifications
Specifications
KLRG1
2F1
Brilliant Violet 421
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Hamster
IgG2, κ
RUO
Primary
Murine
Flow Cytometry
0.2mg/mL
Klrg1; Killer cell lectin-like receptor subfamily G member 1; MAFA
Affinity Purified
A-LAK from C57BL/6 mice
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal