Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KPNA3 Goat anti-Bovine, Canine, Human, Mouse, Rat, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Supplier:  Thermo Scientific PA132096

Catalog No. PIPA132096

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

PA1-32096 detects KPNA3 from human samples. PA1-32096 has been successfully used in Western blot applications. The PA1-32096 immunogen is: Peptide with sequence DPTANLQTKEFNF, from C Terminus of the protein sequence according to NP_002258 Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

The transport of molecules between the nucleus and the cytoplasm in eukaryotic cells is mediated by the nuclear pore complex (NPC), which consists of 60-100 proteins. Small molecules (up to 70 kD) can pass through the nuclear pore by nonselective diffusion while larger molecules are transported by an active process. The protein encoded by this gene belongs to the importin alpha family, and is involved in nuclear protein import.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

KPNA3
Polyclonal
Unconjugated
O00505, O35344
KPNA3
Peptide with sequence DPTANLQTKEFNF, from C Terminus of the protein sequence according to NP_002258
100 μg
Primary
Bovine, Canine, Human, Mouse, Rat
Antibody
IgG
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
KPNA3
RP11-432M24.3, IPOA4, SRP1, SRP1gamma, SRP4, hSRP1
Goat
Antigen affinity chromatography
RUO
16648, 361055, 3839, 476909, 540719
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit