Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KPNA4 Goat anti-Bovine, Canine, Human, Mouse, Rat, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA132097

Catalog No. PA132097


Add to cart

Description

Description

PA1-32097 detects KPNA4 from human, mouse, rat, dog, and cow samples. PA1-32097 has been successfully used in Western blot applications. The PA1-32097 immunogen is a peptide with sequence C-NSSANVPTEGFQF. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

The nuclear import of karyophilic proteins is directed by short amino acid sequences termed nuclear localization signals (NLSs). Karyopherins, or importins, are cytoplasmic proteins that recognize NLSs and dock NLS-containing proteins to the nuclear pore complex. The protein encoded by this gene shares the sequence similarity with Xenopus importin-alpha and Saccharomyces cerevisiae Srp1. This protein is found to interact with the NLSs of DNA helicase Q1 and SV40 T antigen.
Specifications

Specifications

KPNA4
Polyclonal
Unconjugated
KPNA4
Liquid
KPNA4
Peptide with sequence C-NSSANVPTEGFQF.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Bovine, Canine, Human, Mouse, Rat
ELISA, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
O00629, O35343
IPOA3, MGC12217, MGC26703, QIP1, SRP3
Goat
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
16649, 361959, 3840, 478680, 535090
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit