Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRPSAP1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523903

Catalog No. PIPA523903

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse and rat based on sequence homology.

Specifications

Specifications

PRPSAP1
Polyclonal
PBS with 0.09% sodium azide
Q14558
PAP39, 39 kDa phosphoribosypyrophosphate synthase-associated protein, PRPP synthase-associated protein 1, phosphoribosyl pyrophosphate synthase-associated protein 1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5635
Flow Cytometry, Western Blot
Unconjugated
PRPSAP1
Liquid
PRPSAP1
KLH conjugated synthetic peptide between 30-59 amino acids from the N-terminal region of human KPRA
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit