Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRI1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523746

Catalog No. PIPA523746

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene overlaps with the gene for cysteineendopeptidase AUT-like 4 in a head-to-tail orientation.
Specifications

Specifications

KRI1
Polyclonal
PBS with 0.09% sodium azide
Q8N9T8
protein KRI1 homolog
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
65095
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
KRI1
Liquid
KRI1
KLH conjugated synthetic peptide between 12-41 amino acids from the N-terminal region of human KRI1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit