Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRIP-1 Mouse, Unlabeled, Clone: 23, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610680

Catalog No. BDB610680


Add to cart

Description

Description

Several gene regulatory proteins contain “zinc finger” domains of about 30 amino acids and N-terminal regions of about 75 amino acids, named the Krnppel-associated box A (KRAB-A). The KRAB-A domains have been implicated in transcriptional repression. KRIP-1 (KRAB-A-interacting protein 1) was identified as a result of its interaction with a KRAB-A domain. KRIP-1 is 834 amino acids with an apparent molecular weight of 105kDa. Sequence alignments indicate that KRIP-1 shares identity with TIF-1, a transcriptional regulator of nuclear receptors. The region responsible for the transcriptional activity in KRIP-1 is located in the C-terminal region, which also contains a Cys4-His-Cys3 PHD finger domain.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

KRIP-1
23
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
KRAB-A Interacting Protein-1
Affinity Purified
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50μg
Cell Biology
Primary
Canine, Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit