Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 35 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524915

Catalog No. PIPA524915

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

KRT35 is a member of the keratin gene family. This type I hair keratin is an acidic protein which heterodimerizes with type II keratins to form hair and nails. The type I hair keratins are clustered in a region of chromosome 17q12-q21 and have the same direction of transcription.
Specifications

Specifications

Cytokeratin 35
Polyclonal
PBS with 0.09% sodium azide
Q92764
HA5, Ha-5, KRTHA5, hHa5, HHa5 hair keratin type I intermediate filament, K35, hair keratin, type I Ha5, hard keratin, type I, 5, keratin, hair, acidic, 5, keratin, type I cuticular Ha5, keratin-35
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3886
Immunohistochemistry, Western Blot
Unconjugated
KRT35
Liquid
KRT35
KLH conjugated synthetic peptide between 352-380 amino acids from the C-terminal region of human KRT35
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit