Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 4 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA513684

Catalog No. PIPA513684


Add to cart

Description

Description

The protein is a member of the keratin family. The type II cytokeratins consist of basic or neutral proteins which are arranged in pairs of heterotypic keratin chains coexpressed during differentiation of simple and stratified epithelial tissues. This type II cytokeratin is specifically expressed in differentiated layers of the mucosal and esophageal epithelia with family member KRT13.
Specifications

Specifications

Cytokeratin 4
Polyclonal
PBS with 0.09% sodium azide
P19013, P07744
Keratin, type II cytoskeletal 4; Cytokeratin-4; CK-4; Keratin-4; K4; CYK4; KRT4
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
16682, 3851
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
KRT4
Liquid
KRT4
KLH conjugated synthetic peptide between 386-415 amino acids from the C-terminal region of human KRT4
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit