Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT82 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524971

Catalog No. PIPA524971


Add to cart

Description

Description

KRT82 is a member of the keratin gene family. As a type II hair keratin, it is a basic protein which heterodimerizes with type I keratins to form hair and nails. The type II hair keratins are clustered in a region of chromosome 12q13 and are grouped into two distinct subfamilies based on structure similarity. One subfamily, consisting of KRTHB1, KRTHB3, and KRTHB6, is highly related. The other less-related subfamily includes KRTHB2, KRTHB4, and KRTHB5. All hair keratins are expressed in the hair follicle; this keratin appears to be a hair cuticle-specific keratin.
Specifications

Specifications

KRT82
Polyclonal
Unconjugated
KRT82
Liquid
KRT82
KLH conjugated synthetic peptide between 312-341 amino acids from the central region of human KRT82
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q9NSB4, Q99M74
HB2, Hb-2, KRTHB2, K82, hard keratin, type II, 2, keratin, hair, basic, 2, keratin, type II cuticular Hb2, keratin-82, type II hair keratin Hb2, type-II keratin Kb22
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
114566, 3888
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit