Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

keratin 81, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human KRTHB1 protein.

Manufacturer:  Abnova Corporation H00003887B01

Catalog No. 89-005-685


Add to cart

Description

Description

The protein encoded by this gene is a member of the keratin gene family. As a type II hair keratin, it is a basic protein which heterodimerizes with type I keratins to form hair and nails. The type II hair keratins are clustered in a region of chromosome 12q13 and are grouped into two distinct subfamilies based on structure similarity. One subfamily, consisting of KRTHB1, KRTHB3, and KRTHB6, is highly related. The other less-related subfamily includes KRTHB2, KRTHB4, and KRTHB5. All hair keratins are expressed in the hair follicle; this hair keratin, as well as KRTHB3 and KRTHB6, is found primarily in the hair cortex. Mutations in this gene and KRTHB6 have been observed in patients with a rare dominant hair disease, monilethrix. [provided by RefSeq

Sequence: MKATVIRHGETLRRTKEEINELNRMIQRLTAEVENAKCQNSKLEAAVAQSEQQGEAALSDARCKLAELEGALQKAKQDMACLIREYQEVMNSKLGLDIEIATYRRLLEGEEQRLCEGIGAVNVCVSSSRGGVVCGDLCVSGSRPVTGSVCSAPCNGNVAVSTGLCAPCGQLNTTCGGGSCGVGSCGISSLGVGSCGSSCRKC
Specifications

Specifications

keratin 81
Unconjugated
No additive
BC021241.2
HB1|Hb-1|KRTHB1|MLN137|ghHkb1|hHAKB2-1
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
3887
Western Blot
Mouse polyclonal antibody raised against a full-length human KRTHB1 protein.
KRT81
Serum
KRT81
KRTHB1 (AAH21241.1, 1 a.a. ~ 202 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit