Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KT3 Epitope Tag Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11040655
Description
KT3 Epitope Tag Polyclonal antibody specifically detects KT3 Epitope Tag in Epitope Tag samples. It is validated for Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation.Specifications
KT3 Epitope Tag | |
Polyclonal | |
Unconjugated | |
PBS with 0.09% Sodium Azide | |
Rabbit | |
Affinity Purified | |
RUO | |
Store at 4C. Do not freeze. | |
Epitope Tag |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
1.0 mg/mL | |
Western Blot 1:1000-1:10000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:100-1:400, Immunoprecipitation | |
KPPTPPPEPET Epitope Tag, KPPTPPPEPET Tag, KT3 Epitope Tag | |
Rabbits were immunized with KT3 (KPPTPPPEPET) conjugated to KLH. Antibody was isolated by affinity chromatography using the peptide immobilized on solid support. | |
0.1 mL | |
Primary | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction