Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POGLUT1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526681

Catalog No. PIPA526681

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine based on sequence homology.

Specifications

Specifications

POGLUT1
Polyclonal
PBS with 0.09% sodium azide
Q8NBL1
C3orf9, CLP46, KDELCL1, KTELC1, MDSRP, Rumi, hCLP46, 9630046K23Rik, CAP10-like 46 kDa protein, CAP10-like protein, 46 kDa, KDELC family like 1, KTEL (Lys-Tyr-Glu-Leu) containing 1, KTEL motif-containing protein 1, myelodysplastic syndromes relative protein, x 010 protein
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
56983
Western Blot
Unconjugated
POGLUT1
Liquid
POGLUT1
KLH conjugated synthetic peptide between 353-382 amino acids from the C-terminal region of human KTEL1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit