Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KV1.2 (KCNA2) Rabbit anti-Bovine, Canine, Human, Mouse, Porcine, Rat, Xenopus laevis, Zebrafish, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523006

Catalog No. PIPA523006


Add to cart

Description

Description

Suggested positive control: HEK293 cell lysate.

Kcna2 is a voltage-gated potassium channel (KV) that belongs to the 6-TM family of potassium channel and also comprises the Ca2+-activated Slo (actually 7-TM) and the Ca2+-activated SK subfamilies. The alpha-subunits contain a single pore-forming region and combine to form tetramers. Potassium channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. The diverse functions of potassium channels include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. Four sequence-related potassium channel genes - shaker, shaw, shab, and shal - have been identified in Drosophila, and each has been shown to have human homolog(s). The Kcna2 gene encodes a member of the potassium channel, voltage-gated, shaker-related subfamily. Kcna2 contains six membrane-spanning domains with a shaker-type repeat in the fourth segment. Further, Kcna2 belongs to the delayed rectifier class, members of which allow nerve cells to efficiently repolarize following an action potential. The coding region of the Kcna2 gene is intronless, and the gene is clustered with genes KCNA3 and KCNA10 on chromosome 1. Diseases associated with KCNA2 include Epileptic Encephalopathy (Early Infantile, 32) and Undetermined Early-Onset Epileptic Encephalopathy.
Specifications

Specifications

KV1.2 (KCNA2)
Polyclonal
Unconjugated
KCNA2
Liquid
Kcna2, kcna2.S, LOC100537815
Synthetic peptide made to a portion of rat Kv1.2 (within residues 50-100).
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Bovine, Canine, Human, Mouse, Porcine, Rat, Xenopus laevis, Zebrafish
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
1 mg/mL
PBS with 0.1% sodium azide
P63142, Q28293, P16389, P63141
RP11-284N8.1, HBK5, HK4, HUKIV, KV1.2, MK2, NGK1, RBK2, potassium voltage-gated channel subfamily A member 2, voltage-gated K(+) channel HuKIV, voltage-gated potassium channel HBK5, voltage-gated potassium channel protein Kv1.2, voltage-gated potassium channel subunit Kv1.2
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
100537815, 16490, 25468, 3737, 373840, 397292, 404022, 539608
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit