Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Kv1, Biotin, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP142788B

Catalog No. NBP142788B


Add to cart

Description

Description

Kv1 Polyclonal antibody specifically detects Kv1 in Human, Mouse, Rat, Porcine, Bovine, Canine, Xenopus, Zebrafish samples. It is validated for Western Blot.
Specifications

Specifications

Kv1
Biotin
PBS with 0.05% Sodium Azide
potassium channel, shaker-related subfamily, voltage-gated potassium channel
Rabbit
IgG
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
Reacts with all members of the Kv1 family. Does not cross-react with Kv3.1.
Western Blot
Western Blot
Affinity Purified
KCNA2
Synthetic peptide made to rat Kv1.2 (within residues 50-100), which is identical in all members of the Kv1 family.
80 kDa
0.1 ml
Lipid and Metabolism, Neuroscience, Neurotransmission, Potassium Channels
Primary
3737
Bovine, Canine, Human, Mouse, Porcine, Rat, Xenopus, Zebrafish
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only