Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kv10.2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP184934
Description
Kv10.2 Polyclonal specifically detects Kv10.2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Kv10.2 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
hEAG2, potassium channel HEAG2, potassium voltage-gated channel, subfamily H (eag-related), member 5, voltage-gated potassium channel subunit Kv10.2 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
KCNH5 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:GDYEVIDEVTNTIQIDSWLYQLALSIGTPYRYNTSAGIWEGGPSKDS | |
0.1 mL | |
Apoptosis, Cancer, DNA Double Strand Break Repair, Protein Phosphatase, Signal Transduction, Tumor Suppressors | |
27133 | |
Human | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction