Learn More
anti-Kv12.2, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP130641
Catalog No. NBP130641
Add to cart
Description
Description
Kv12.2 Polyclonal antibody specifically detects Kv12.2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
Kv12.2
LYOPH
Western Blot 1:300-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Antisera
KCNH3
A synthetic peptide from aa region 100-150 of human KCNH3 conjugated to blue carrier protein was used as the antigen.
Unpurified
RUO
Primary
23416
Reconstitute with 0.1 ml sterilized water to desired concentration.
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Whole antisera with No Preservative
BEC1, brain-specific eag-like channel 1, ELK channel 2, ELK2, ether-a-go-go K(+) channel family member, ether-a-go-go-like potassium channel 2, KIAA1282, Kv12.2, potassium voltage-gated channel subfamily H member 3, potassium voltage-gated channel, subfamily H (eag-related), member 3, voltage-gated potassium channel subunit Kv12.2
Rabbit
IgG
0.1 ml
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Appears to be specific for KCNH3.
Human